Ethyl-3-methylcrotonate 98%

Supplier: Ambeed
Danger

Synonyms: 3,3-Dimethylacrylic acid ethyl ester, Ethyl 3,3-Dimethylacrylate, Ethyl 3-methylbut-2-enoate, 3-Methyl-2-butenoic acid ethyl ester, 3-Methylcrotonic acid ethyl ester, Ethyl 3-Methyl-2-butenoate

A466900-25G A466900-100G
76804-722EA 49 USD
76804-722 77594-472
Ethyl-3-methylcrotonate 98%
Ethyl-3-methylcrotonate

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
MDL Number: MFCD00009187
CAS Number: 638-10-8
UN: 3272
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR