9-Fluorenylmethyl pentafluorophenyl carbonate 97%

Supplier: Ambeed
Warning

Synonyms: Carbonic Acid 9-Fluorenylmethyl Pentafluorophenyl Ester, Fmoc-OPfp, 9-Fluorenylmethyl Pentafluorophenyl Carbonate

A465503-1G A465503-250MG A465503-5G
76804-712EA 35.01 USD
76804-712 76804-714 76804-716
9-Fluorenylmethyl pentafluorophenyl carbonate 97%
9-Fluorenylmethyl pentafluorophenyl carbonate

Precaución: for Research Use Only

Formula: C₂₁H₁₁F₅O₃
MW: 406.31 g/mol
MDL Number: MFCD00013261
CAS Number: 88744-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR