3-(4-Phenylpiperazin-1-yl)propane-1,2-diol 98% (by NMR)

Supplier: Ambeed
Warning

A464373-100G A464373-100MG A464373-1G A464373-25G A464373-25MG A464373-500G A464373-5G
76804-692EA 88 USD
76804-692 76804-694 76804-696 76804-698 76804-700 76804-702 76804-704
3-(4-Phenylpiperazin-1-yl)propane-1,2-diol 98% (by NMR)
3-(4-Phenylpiperazin-1-yl)propane-1,2-diol

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₂
MDL Number: MFCD00079124
CAS Number: 17692-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR