Ethyl-10-bromodecanoate 96%

Supplier: Ambeed
Warning

A463542-100G A463542-100MG A463542-1G A463542-250MG A463542-25G A463542-5G
76804-680EA 604 USD
76804-680 76804-682 76804-684 76804-686 76804-688 76804-690
Ethyl-10-bromodecanoate 96%
Ethyl-10-bromodecanoate

Precaución: for Research Use Only

Formula: C₁₂H₂₃BrO₂
MDL Number: MFCD01112172
CAS Number: 55099-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR