Fmoc-2,4-dichloro-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(2,4-DiCl)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid, 2,4-Dichloro-L-phenylalanine, N-FMOC protected

A462849-1G A462849-250MG A462849-5G A462849-25G
76804-674EA 12 USD
76804-674 76804-676 76804-678 77880-011
Fmoc-2,4-dichloro-L-phenylalanine 95%
Fmoc-2,4-dichloro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₁₉Cl₂NO₄
MW: 456.33 g/mol
MDL Number: MFCD01863048
CAS Number: 352351-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR