Ethyl decanoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester

A462675-100G A462675-25G A462675-500G
76804-668EA 8.49 USD
76804-668 76804-670 76804-672
Ethyl decanoate 98%
Ethyl decanoate

Precaución: for Research Use Only

Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
MDL Number: MFCD00009581
CAS Number: 110-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR