Ethyl-3-bromobenzoate 95%

Supplier: Ambeed
Warning

A462527-100G A462527-1G A462527-25G A462527-500G A462527-5G
76804-658EA 78 USD
76804-658 76804-660 76804-662 76804-664 76804-666
Ethyl-3-bromobenzoate 95%
Ethyl-3-bromobenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD00013529
CAS Number: 24398-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR