Hexadecyl methacrylate 98+% stabilised
Supplier: Ambeed
|
Danger
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A462464-1G
A462464-25G
A462464-5G
A462464-100G
76804-650EA
14
USD
76804-650
76804-652
76804-654
77880-010
Hexadecyl methacrylate 98+% stabilised
Hexadecyl methacrylate
Precaución: for Research Use Only
Formula:
C₂₀H₃₈O₂ MW: 310.51 g/mol |
MDL Number:
MFCD00078346 CAS Number: 2495-27-4 UN: 3082 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...