Guanabenz Acetate ≥99%

Supplier: Ambeed
Danger

Synonyms: 2-(2,6-Dichlorobenzylidene)hydrazine-1-carboximidamide acetate

A461798-50MG A461798-250MG A461798-5G
76804-646EA 9.55 USD
76804-646 76804-648 77669-078 77880-007
Guanabenz Acetate ≥99%
Guanabenz Acetate

Precaución: for Research Use Only

Formula: C₈H₈Cl₂N₄·C₂H₄O₂
MDL Number: MFCD00153801
CAS Number: 23256-50-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR