Manganese(II) bromide 98%
Supplier: Ambeed
|
Warning
|
A461031-100G
A461031-1G
A461031-25G
A461031-5G
76804-634EA
464
USD
76804-634
76804-636
76804-638
76804-640
Manganese(II) bromide 98%
Manganese(II) bromide
Precaución: for Research Use Only
Formula:
Br₂Mn |
MDL Number:
MFCD00011113 CAS Number: 13446-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...