Imidazo[1,2-a]pyridine-3-carbonitrile 96%

Supplier: Ambeed
Warning

A460106-1G A460106-250MG A460106-5G
76804-622EA 82 USD
76804-622 76804-624 76804-626
Imidazo[1,2-a]pyridine-3-carbonitrile 96%
Imidazo[1,2-a]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₅N₃
MDL Number: MFCD01720448
CAS Number: 6200-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR