Dichlorobis(tributylphosphine)nickel(II) 98%

Supplier: Ambeed
Danger

A458883-1G A458883-250MG A458883-25G A458883-5G
76804-608EA 26 USD
76804-608 76804-610 76804-612 76804-614
Dichlorobis(tributylphosphine)nickel(II) 98%
Dichlorobis(tributylphosphine)nickel(II)

Precaución: for Research Use Only

Formula: C₂₄H₅₄Cl₂NiP₂
MW: 534.23 g/mol
MDL Number: MFCD00049260
CAS Number: 15274-43-8
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR