Glabridin 97%

Supplier: Ambeed
Warning

Synonyms: (R)-4-(8,8-Dimethyl-2,3,4,8-tetrahydropyrano[2,3-f]chromen-3-yl)benzene-1,3-diol

A458176-10MG A458176-1G A458176-1MG A458176-250MG A458176-25MG A458176-5MG A458176-100MG A458176-5G
76804-596EA 8.49 USD
76804-596 76804-598 76804-600 76804-602 76804-604 76804-606 77601-978 77601-980
Glabridin 97%
Glabridin

Precaución: for Research Use Only

Formula: C₂₀H₂₀O₄
MW: 324.37 g/mol
MDL Number: MFCD03427694
CAS Number: 59870-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR