Hexyl salicylate 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Hexyl 2-hydroxybenzoate, N-Hexyl salicylate
A457443-1000G
A457443-100G
A457443-500G
76804-590EA
72
USD
76804-590
76804-592
76804-594
Hexyl salicylate 99+%
Hexyl salicylate
Precaución: for Research Use Only
Formula:
C₁₃H₁₈O₃ |
MDL Number:
MFCD00036487 CAS Number: 6259-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...