Dichlorobis(pyridine)palladium(II) 98%

Supplier: Ambeed
Warning

A456807-100MG A456807-1G A456807-250MG
76804-572EA 64.72 USD
76804-572 76804-574 76804-576
Dichlorobis(pyridine)palladium(II) 98%
Dichlorobis(pyridine)palladium(II)

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂N₂Pd
MW: 335.53 g/mol
MDL Number: MFCD00792248
CAS Number: 14872-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR