Dodecyl acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Lauryl acetate, Acetic acid Lauryl ester, Acetic acid Dodecyl ester
A456295-5G
A456295-100G
A456295-500G
76804-570EA
8
USD
76804-570
77626-902
77626-904
Dodecyl acetate 98%
Dodecyl acetate
Precaución: for Research Use Only
Formula:
C₁₄H₂₈O₂ MW: 228.38 g/mol Boiling Pt: 265 |
MDL Number:
MFCD00008973 CAS Number: 112-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...