Diethylphenylphosphine ≥98%

Supplier: Ambeed
Warning

Synonyms: Diethyl(phenyl)phosphane

A455521-1G A455521-250MG
76804-566EA 39.26 USD
76804-566 76804-568
Diethylphenylphosphine ≥98%
Diethylphenylphosphine

Precaución: for Research Use Only

Formula: C₁₀H₁₅P
MW: 166.21 g/mol
MDL Number: MFCD00015172
CAS Number: 1605-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR