Diethylene glycol monobutyl ether 99+%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Butoxyethoxy)ethanol, Diethylene glycol mono-n-butyl ether, Butyldigol

A455309-100G A455309-500G
76804-562EA 5 USD
76804-562 76804-564
Diethylene glycol monobutyl ether 99+%
Diethylene glycol monobutyl ether

Precaución: for Research Use Only

Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MW: 162.23 g/mol
Boiling Pt: 230.4
Melting Pt: -68
Flash Pt: 100
MDL Number: MFCD00002881
CAS Number: 112-34-5
Merck Index: 13,01556

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR