Ethyl-3-aminobenzofuran-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-Aminocoumarilate, 3-Aminobenzofuran-2-carboxylic acid Ethyl Ester, 3-Aminocoumarilic acid Ethyl Ester, Ethyl 3-aminobenzofuran-2-carboxylate

A455030-100MG A455030-250MG
76804-558EA 21.22 USD
76804-558 76804-560
Ethyl-3-aminobenzofuran-2-carboxylate 98%
Ethyl-3-aminobenzofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
MDL Number: MFCD00641330
CAS Number: 39786-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR