Isobutyl decanoate 98%

Supplier: Ambeed
Warning

A455001-100G A455001-25G A455001-5G
76804-552EA 129 USD
76804-552 76804-554 76804-556
Isobutyl decanoate 98%
Isobutyl decanoate

Precaución: for Research Use Only

Formula: C₁₄H₂₈O₂
MW: 228.3700 g/mol
MDL Number: MFCD00059259
CAS Number: 30673-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR