L-Norleucinol 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Aminohexan-1-ol, (S)-(+)-2-Amino-1-hexanol

A454381-100MG A454381-1G A454381-250MG A454381-5G
76804-544EA 58.35 USD
76804-544 76804-546 76804-548 77601-958
L-Norleucinol 95%
L-Norleucinol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
MW: 117.189 g/mol
MDL Number: MFCD02683226
CAS Number: 80696-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR