Triethylene Glycol Bis(p-toluenesulfonate) 97%
Supplier: Ambeed
|
Warning
|
A452679-100G
A452679-10G
A452679-1G
A452679-25G
A452679-500G
A452679-5G
A452679-1000G
76804-528EA
59
USD
76804-528
76804-530
76804-532
76804-534
76804-536
77626-896
77879-990
Triethylene Glycol Bis(p-toluenesulfonate) 97%
Triethylene Glycol Bis(p-toluenesulfonate)
Precaución: for Research Use Only
Formula:
C₂₀H₂₆O₈S₂ |
MDL Number:
MFCD00048096 CAS Number: 19249-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...