meso-Butane-1,2,3,4-tetracarboxylic acid 98+%

Supplier: Ambeed
Warning

Synonyms: trans-Butane-1,2,3,4-tetracarboxylic acid

A451074-100G A451074-25G
76804-522EA 72.15 USD
76804-522 76804-524
meso-Butane-1,2,3,4-tetracarboxylic acid 98+%
meso-Butane-1,2,3,4-tetracarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₀O₈
MW: 234.16 g/mol
MDL Number: MFCD00070496
CAS Number: 4534-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR