Dimidium bromide 98%

Supplier: Ambeed
Warning

Synonyms: 2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide

A449281-100MG A449281-1G A449281-250MG A449281-5G
76804-498EA 22.28 USD
76804-498 76804-500 76804-502 76804-504
Dimidium bromide 98%
Dimidium bromide

Precaución: for Research Use Only

Formula: C₂₀H₁₈BrN₃
MW: 380.29 g/mol
MDL Number: MFCD00011757
CAS Number: 518-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR