2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Boc-thio-4,6-dimethylpyrimidine, O-tert-Butyl S-(4,6-dimethylpyrimidin-2-yl) carbonothioate

A448177-25G A448177-5G A448177-1G
76804-486EA 137.92 USD
76804-486 76804-488 77669-012
2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine 95%
2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₂S
MW: 240.32 g/mol
MDL Number: MFCD00006080
CAS Number: 41840-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR