L-Isoglutamine 98%

Supplier: Ambeed
Warning

Synonyms: H-Glu-NH₂, (S)-4,5-Diamino-5-oxopentanoic acid

A447862-1G A447862-250MG A447862-5G
76804-480EA 103.97 USD
76804-480 76804-482 76804-484
L-Isoglutamine 98%
L-Isoglutamine

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O₃
MW: 146.146 g/mol
MDL Number: MFCD00135661
CAS Number: 636-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR