Lomustine 95%

Supplier: Ambeed
Danger

A447678-250MG A447678-50MG
76804-474EA 5.31 USD
76804-474 76804-476 76804-478
Lomustine 95%
Lomustine

Precaución: for Research Use Only

Formula: C₉H₁₆ClN₃O₂
MDL Number: MFCD00012392
CAS Number: 13010-47-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR