(1S,4aS,7aS)-7-(Hydroxymethyl)-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A447520-10MG A447520-1MG A447520-50MG A447520-5MG
76804-466EA 20.16 USD
76804-466 76804-468 76804-470 76804-472
(1S,4aS,7aS)-7-(Hydroxymethyl)-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylic acid 98%
(1S,4aS,7aS)-7-(Hydroxymethyl)-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₁₀
MW: 374.33 g/mol
MDL Number: MFCD00210294
CAS Number: 27741-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR