1,2-Diphenyl-1,1,2,2-tetramethyldisilane 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,1,2,2-Tetramethyl-1,2-diphenyldisilane, 1,1,2,2-Tetramethyl-1,2-diphenyldisilane, 1,2-Diphenyltetramethyldisilane
A446320-1G
A446320-25G
A446320-5G
76804-452EA
26.53
USD
76804-452
76804-454
77572-398
1,2-Diphenyl-1,1,2,2-tetramethyldisilane 97%
1,2-Diphenyl-1,1,2,2-tetramethyldisilane
Precaución: for Research Use Only
Formula:
C₁₆H₂₂Si₂ MW: 270.52 g/mol |
MDL Number:
MFCD00054786 CAS Number: 1145-98-8 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...