3-Morpholinobenzaldehyde 98%

Supplier: Ambeed
Warning

A446045-100MG A446045-1G A446045-250MG A446045-25G A446045-5G
76804-432EA 27.59 USD
76804-432 76804-434 76804-436 76804-438 76804-440
3-Morpholinobenzaldehyde 98%
3-Morpholinobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.2264 g/mol
MDL Number: MFCD07772811
CAS Number: 446866-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR