4-[4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy]butyric acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-(4-(1-Hydroxyethyl)-2-methoxy-5-nitro-phenoxy)butyric acid, 4-(4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy)butanoic acid

A444508-100MG A444508-1G A444508-250MG A444508-5G
76804-410EA 74.27 USD
76804-410 76804-412 76804-414 76804-416
4-[4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy]butyric acid 95%
4-[4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy]butyric acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₇
MW: 299.279 g/mol
MDL Number: MFCD01318869
CAS Number: 175281-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR