2-Amino-2-cyclopropylethan-1-ol hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-2-cyclopropylethanol hydrochloride

A444168-250MG A444168-100MG
76804-408EA 250.38 USD
76804-408 77668-988
2-Amino-2-cyclopropylethan-1-ol hydrochloride 95%
2-Amino-2-cyclopropylethan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO·ClH
MW: 137.608 g/mol
MDL Number: MFCD18839092
CAS Number: 1306603-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR