Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) 95%
Supplier: Ambeed
|
Warning
|
A443379-1000G
A443379-100G
A443379-25G
A443379-500G
76804-400EA
19
USD
76804-400
76804-402
76804-404
76804-406
Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) 95%
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate)
Precaución: for Research Use Only
Formula:
C₂H₅NaO₄S |
MDL Number:
MFCD00007534 CAS Number: 1562-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...