Dibenzyl sulphoxide 99%

Supplier: Ambeed
Warning

Synonyms: Benzyl sulphoxide, Dibenzyl sulphoxide

A442496-100G A442496-25G A442496-5G
76804-388EA 69 USD
76804-388 76804-390 76804-392
Dibenzyl sulphoxide 99%
Dibenzyl sulphoxide

Precaución: for Research Use Only

Formula: C₁₄H₁₄OS
MDL Number: MFCD00004782
CAS Number: 621-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR