FMOC-Hmd HCl 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(9H-Fluoren-9-yl)methyl (6-aminohexyl)carbamate hydrochloride
A441360-1G
A441360-250MG
A441360-5G
76804-384EA
73
USD
76804-384
76804-386
77879-971
FMOC-Hmd HCl 95%
FMOC-Hmd HCl
Precaución: for Research Use Only
Formula:
C₂₁H₂₆N₂O₂·ClH |
MDL Number:
MFCD00830743 CAS Number: 945923-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...