5,7-Dihydroxy-6,8-dimethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one 99+%

Supplier: Ambeed
Warning

A440977-1MG A440977-25MG A440977-5MG
76804-378EA 8 USD
76804-378 76804-380 76804-382
5,7-Dihydroxy-6,8-dimethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one 99+%
5,7-Dihydroxy-6,8-dimethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₈H₁₆O₇
MDL Number: MFCD08689947
CAS Number: 10176-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR