DL-4-Hydroxyphenyllactic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-3-(4-hydroxyphenyl)propanoic acid, Hydroxyphenyllactic acid, 2-Hydroxy-3-(4-hydroxyphenyl)propionic Acid

A440400-100MG A440400-1G A440400-250MG A440400-50MG A440400-5G
76804-370EA 47 USD
76804-370 76804-372 76804-374 76804-376 77879-969
DL-4-Hydroxyphenyllactic acid 98%
DL-4-Hydroxyphenyllactic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MDL Number: MFCD00010514
CAS Number: 306-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR