4-(4-Thiomorpholinylcarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Thiomorpholine-4-carbonyl)phenylboronic acid, (4-(Thiomorpholine-4-carbonyl)phenyl)boronic acid

A440331-100MG A440331-1G A440331-250MG A440331-5G
76804-362EA 36.08 USD
76804-362 76804-364 76804-366 76804-368
4-(4-Thiomorpholinylcarbonyl)phenylboronic acid 98%
4-(4-Thiomorpholinylcarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄BNO₃S
MW: 251.1100 g/mol
MDL Number: MFCD06659879
CAS Number: 850568-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR