Ethyl 4-(chlorosulfonyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%

Supplier: Ambeed
Danger

Synonyms: 2,4-Dimethyl-5-(ethoxycarbonyl)-1H-pyrrole-3-sulphonyl chloride, 3-(Chlorosulphonyl)-2,4-dimethyl-5-(ethoxycarbonyl)-1H-pyrrole

A440090-100MG A440090-1G A440090-250MG
76804-356EA 76.39 USD
76804-356 76804-358 76804-360
Ethyl 4-(chlorosulfonyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%
Ethyl 4-(chlorosulfonyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₄S
MW: 265.714 g/mol
MDL Number: MFCD03407326
CAS Number: 368869-88-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR