Ethyl-2-mercaptopropionate 97%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-mercaptopropanoate, 2-Mercaptopropionic Acid Ethyl Ester

A438323-25G A438323-5G
76804-348EA 78.51 USD
76804-348 76804-350
Ethyl-2-mercaptopropionate 97%
Ethyl-2-mercaptopropionate

Precaución: for Research Use Only

Formula: C₅H₁₀O₂S
MW: 134.19 g/mol
MDL Number: MFCD00040233
CAS Number: 19788-49-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR