Furfuryl glycidyl ether 97%

Supplier: Ambeed
Warning

Synonyms: 2-((Oxiran-2-ylmethoxy)methyl)furan

A437502-100G A437502-1G A437502-25G A437502-5G
76804-338EA 221.73 USD
76804-338 76804-340 76804-342 76804-344
Furfuryl glycidyl ether 97%
Furfuryl glycidyl ether

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MW: 154.165 g/mol
MDL Number: MFCD00013348
CAS Number: 5380-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR