Fmoc-D-3-(4-thiazolyl)-Alanine 96%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiazol-4-yl)propanoic acid

A436534-1G A436534-250MG A436534-5G
76804-332EA 75 USD
76804-332 76804-334 76804-336
Fmoc-D-3-(4-thiazolyl)-Alanine 96%
Fmoc-D-3-(4-thiazolyl)-Alanine

Precaución: for Research Use Only

Formula: C₂₁H₁₈N₂O₄S
MDL Number: MFCD00672569
CAS Number: 205528-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR