Ethyl-2-chloro-6-hydroxybenzoate 98%

Supplier: Ambeed
Warning

A434153-1G A434153-250MG A434153-5G
76804-320EA 42 USD
76804-320 76804-322 76804-324
Ethyl-2-chloro-6-hydroxybenzoate 98%
Ethyl-2-chloro-6-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD09038111
CAS Number: 112270-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR