4-Chloro-1-naphthol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloronaphthalen-1-ol, 1-Chloro-4-hydroxynaphthalene, 4-CN, Chloronaphthol
A431158-1G
A431158-5G
A431158-25G
A431158-10G
76804-288EA
17
USD
76804-288
76804-290
77879-949
77879-948
4-Chloro-1-naphthol 97%
4-Chloro-1-naphthol
Precaución: for Research Use Only
Formula:
ClC₁₀H₆OH |
MDL Number:
MFCD00003974 CAS Number: 604-44-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...