Methyl-2,6-dichlorobenzoate 98%

Supplier: Ambeed
Warning

A430092-5G A430092-10G A430092-1G A430092-25G
76804-274EA 41 USD
76804-274 77601-852 77601-854 77601-856
Methyl-2,6-dichlorobenzoate 98%
Methyl-2,6-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
MDL Number: MFCD01313701
CAS Number: 14920-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR