Fmoc-D-Tyr(3-I)-OH 95%
Supplier: Ambeed
|
Warning
|
A428369-1G
A428369-5G
A428369-10G
76804-260EA
37
USD
76804-260
76804-262
77879-938
Fmoc-D-Tyr(3-I)-OH 95%
Fmoc-D-Tyr(3-I)-OH
Precaución: for Research Use Only
Formula:
C₂₄H₂₀INO₅ MW: 529.3 g/mol |
MDL Number:
MFCD03840398 CAS Number: 244028-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...