4-(5,6,7,8-Tetrahydroimidazo[1,5-a]pyridin-5-yl)benzonitrile 95%

Supplier: Ambeed
Danger

A428094-10MG A428094-1MG A428094-5MG
76804-248EA 78.51 USD
76804-248 76804-250 76804-252
4-(5,6,7,8-Tetrahydroimidazo[1,5-a]pyridin-5-yl)benzonitrile 95%
4-(5,6,7,8-Tetrahydroimidazo[1,5-a]pyridin-5-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₃N₃
MW: 223.27 g/mol
MDL Number: MFCD02313480
CAS Number: 102676-47-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR