Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A427139-10G A427139-1G A427139-250MG A427139-25G A427139-5G
76804-232EA 151 USD
76804-232 76804-234 76804-236 76804-238 76804-240
Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate 95%
Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MDL Number: MFCD00159638
CAS Number: 35691-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR