Ethyl 3,4-Bis(2-methoxyethoxy)benzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,4-Bis(2-methoxyethoxy)benzoic acid Ethyl ester, Ethyl 3,4-bis(2-methoxyethoxy)benzoate
A426855-100G
A426855-10G
A426855-1G
A426855-25G
A426855-5G
76804-222EA
184.6
USD
76804-222
76804-224
76804-226
76804-228
76804-230
Ethyl 3,4-Bis(2-methoxyethoxy)benzoate 98%
Ethyl 3,4-Bis(2-methoxyethoxy)benzoate
Precaución: for Research Use Only
Formula:
C₁₅H₂₂O₆ MW: 298.34 g/mol |
MDL Number:
MFCD01935284 CAS Number: 183322-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...