H-α-Me-DL-Val-Oh 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-α-Me-Val-OH, 2-Amino-2,3-dimethylbutanoic acid, H-α-Me-DL-Val-OH

A426838-1G A426838-5G
76804-218EA 22 USD
76804-218 76804-220
H-α-Me-DL-Val-Oh 97%
H-alpha-Me-DL-Val-Oh

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.175 g/mol
MDL Number: MFCD00672376
CAS Number: 26287-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR